Acetonitrile, low water content ≥99.9% (by GC), BioSyn™ for biosynthesis, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Methanecarbonitrile, Methane, cyano-, Methyl cyanide, Ethyl nitrile, ACN, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

BC017-1250 BC017-200 BC017-56
BJBC017-1250EA 30908.2 USD
BJBC017-1250 BJBC017-200 BJBC017-56
Acetonitrile, low water content ≥99.9% (by GC), BioSyn™ for biosynthesis, Burdick & Jackson™
Acetonitrile
Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6
Melting Pt: -45.7
Density: 0.7822 g/cm³
Flash Pt: 6 °C
MDL Number: MFCD00001878
CAS Number: 75-05-8
UN: 1648
ADR: 3 II
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GC Analysis 99.9% min
Refractive Index (20 °C) 1.3434 - 1.3446
Residue Max. 1 mg/l
UV Absorbance at 200 nm Max. 0.050 AU
UV Absorbance at 210 nm Max. 0.040 AU
UV Absorbance at 225 nm Max. 0.010 AU
UV Cutoff Max. 190 nm
Water by Karl Fischer Titration Max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR