Pyridine ≥99.8%, BioSyn™ for biosynthesis, for applications requiring low water content, Burdick & Jackson™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Azabenzene, Azine
BB333-4
BC333-200
BB333-2.5L
BJBB333-4CS
5055.35
USD
BJBB333-4
BJBC333-200
BJBN333-20
BJBB333-2.5L
Pyridine ≥99.8%, BioSyn™ for biosynthesis, for applications requiring low water content, Burdick & Jackson™
Pyridine
BioSyn« products are ideal for DNA, RNA and oligonucleotide synthesis. All solvents and regents are filtered (0.2-micron) and tested for oxidation reage.
- Ultra-low water specifications
- Low amine specifications
- Low particulate levels
- High solvent purity
Formula:
C₅H₅N MW: 79.1 g/mol Boiling Pt: 12.4 %(V) Melting Pt: –41.55 °C Density: 0.983 g/cm³ (20 °C) Flash Pt: 20 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011732 CAS Number: 110-86-1 EINECS: 203-809-9 UN: 1282 ADR: 3,II Merck Index: 13,08060 |
|
Specification Test Results
Amines by Ninhydrin Test | 10 ppm max. |
Free Acid | Passes test |
GC Analysis | 99.8 % min. |
Refractive Index (20ºC) | 1.5085 - 1.5109 |
Residue | 1 mg/L max. |
Water by Karl Fischer Titration | 50 ppm max. |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...