Methanol ≥99.9%, BioSyn® for biosynthesis, for applications requiring low water content, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger


Filtered through a 0.2 µm filter.

BB233-4 BC233-200 BC233-56
BJBB233-4CS 765 USD
BJBB233-4 BJBC233-200 BJBC233-56
Methanol ≥99.9%, BioSyn® for biosynthesis, for applications requiring low water content, Burdick & Jackson™
Methanol

BioSyn™ products are ideal for DNA, RNA and oligonucleotide synthesis.


  • Ultra-low water specifications
  • Low amine specifications
  • Low particulate levels
  • High solvent purity

Formula: CH₃OH
Boiling Pt: 64.7 °C
Melting Pt: –98 °C
Density: 0.792 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GC Analysis 99.9 % min.
Refractive Index (20ºC) 1.3280 - 1.3288
Residue 1 mg/L max.
UV Absorbance @ 225 nm 0.16 AU max.
UV Absorbance @ 250 nm 0.02 AU max.
UV Absorbance @ 300 nm 0.005 AU max.
UV Absorbance @ 400 nm 0.005 AU max.
UV Cutoff 205 nm max.
Water by Karl Fischer Titration 300 ppm max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR