METHANOL 4L
Supplier: Honeywell Research Chemicals
Methyl alcohol. CAS RN 67-56-1. Formula Weight: 32.04. Density: 0.79kg/L. B&J ACS/HPLC, 99.9% min. by GC. For HPLC, spectrophotometry and applications requiring ACS Reagent Grade solvents. Subject to postal/shipping restrictions. 4L.