Chloroform ≥99.9% stabilised ACS, meets analytical specification of USP for HPLC, for spectrophotometry

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

Stabilization: Stabilised with amylene

AH049-4 AH049-4PC
BJAH049-4CS 1469 USD
BJAH049-4 BJAH049-4PC
Chloroform ≥99.9% stabilised ACS, meets analytical specification of USP for HPLC, for spectrophotometry
Chloroform

For HPLC, spectrophotometry, and applications requiring ACS reagent grade solvents. Stabilized with amylene. UV cutoff at 245nm.


  • ACS certified
  • Lot-to-lot consistency
  • HPLC grade controlled for UV absorbance, GC assay, water content and non-volatile residue

Formula: CHCl₃
Boiling Pt: 62 °C
Melting Pt: –64 °C
Density: 1.49 g/cm³ (20 °C)
Flash Pt: Does not flash
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Acetone and Aldehyde Passes test
Water by Karl Fischer Titration 0.02 % max.
Acid and Chloride Passes test
Color (APHA) 10 max.
Free Chlorine Passes test
GC Analysis 99.9 % min.
Lead 0.05 ppm max.
Refractive Index (20ºC) 1.4454 - 1.4464
Residue 3 mg/L max.
UV Cutoff 245 nm max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR