Acetonitrile ≥99.9% ACS, meets analytical specification of USP for HPLC, for spectrophotometry

Supplier: Honeywell Research Chemicals
Danger

AH015-4 AH015-4PC
BJAH015-4CS 2741.09 USD
BJAH015-4 BJAH015-4PC
Acetonitrile ≥99.9% ACS, meets analytical specification of USP for HPLC, for spectrophotometry
Acetonitrile

Meets USP Reagent Specifications


  • ACS certified
  • Lot-to-lot consistency
  • HPLC grade controlled for UV absorbance, GC assay, water content and non-volatile residue

Formula: H₃CCN
Boiling Pt: 82 °C (1.013 hPa)
Melting Pt: –46 °C
Density: 0.7822 g/cm³ (20 °C)
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Color (APHA) 10 max.
Water by Karl Fisher Titration 0.05 % max.
GC Analysis 99.9 % min.
HPLC Gradient @ 254 nm 0.005 AU max.
Refractive Index (20ºC) 1.3434 - 1.3446
Residue 3 mg/L max.
Titrable Acid 8 µeq/g max.
Titrable Base 0.6 µeq/g max.
UV Absorbance @ 200 nm 0.07 AU max.
UV Cutoff 190 nm max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR