Ammonium heptamolybdate tetrahydrate 81.0-83.0% (MoO₃ basis) ACS, Fluka™

Supplier: Honeywell Research Chemicals

A7302-100G A7302-1KG A7302-500G A7302-6X500G
BJA7302-100GEA 99.69 USD
BJA7302-100G BJA7302-1KG BJA7302-2.5KG BJA7302-500G BJA7302-6X500G
Ammonium heptamolybdate tetrahydrate 81.0-83.0% (MoO₃ basis) ACS, Fluka™
Ammonium heptamolybdate tetrahydrate
Formula: (NH₄)₆Mo₇O₂₄·4H₂O
Density: 2.498 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167059
CAS Number: 12054-85-2
EINECS: 234-320-9

Order Now

Specification Test Results

Assay (as MoO₃) 81.0 - 83.0 %
Water insoluble matter Max. 0.005 %
Potassium (K) Max. 0.01 %
Magnesium (Mg) Max. 0.005 %
Sodium (Na) Max. 0.01 %
Heavy metals (ICP) ACS Max. 0.001 %
Chloride (Cl) Max. 0.002 %
Nitrate (NO₃) Max. 0.003 %
Phosphate, Arsenate, Silicate (as SiO₂) Max. 0.001 %
Phosphate (PO₄) Max. 5 ppm
Sulfate (SO₄) Max. 0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR