Ammonium heptamolybdate tetrahydrate 81.0-83.0% (MoO₃ basis) ACS, Fluka™
Supplier: Honeywell Research Chemicals
A7302-100G
A7302-1KG
A7302-500G
A7302-6X500G
BJA7302-100GEA
99.69
USD
BJA7302-100G
BJA7302-1KG
BJA7302-2.5KG
BJA7302-500G
BJA7302-6X500G
Ammonium heptamolybdate tetrahydrate 81.0-83.0% (MoO₃ basis) ACS, Fluka™
Ammonium heptamolybdate tetrahydrate
Formula:
(NH₄)₆Mo₇O₂₄·4H₂O Density: 2.498 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00167059 CAS Number: 12054-85-2 EINECS: 234-320-9 |
Specification Test Results
Assay (as MoO₃) | 81.0 - 83.0 % |
Water insoluble matter | Max. 0.005 % |
Potassium (K) | Max. 0.01 % |
Magnesium (Mg) | Max. 0.005 % |
Sodium (Na) | Max. 0.01 % |
Heavy metals (ICP) ACS | Max. 0.001 % |
Chloride (Cl) | Max. 0.002 % |
Nitrate (NO₃) | Max. 0.003 % |
Phosphate, Arsenate, Silicate (as SiO₂) | Max. 0.001 % |
Phosphate (PO₄) | Max. 5 ppm |
Sulfate (SO₄) | Max. 0.02 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...