Sodium formate solution 10 mM NaOH in 2-propanol/water (1:1) (+0.2% HCOOH) for LC-MS, Fluka™

Supplier: Honeywell Research Chemicals
Danger

97574-100ML
BJ97574-100MLEA 357 USD
BJ97574-100ML
Sodium formate solution 10 mM NaOH in 2-propanol/water (1:1) (+0.2% HCOOH) for LC-MS, Fluka™
Sodium formate
Formula: HCO₂Na
Flash Pt: 22 °C
Storage Temperature: Ambient
MDL Number: MFCD00013101
CAS Number: 141-53-7
UN: 1219
ADR: 3,II

Order Now

Specification Test Results

Identity (LC-MS) complying
LC-MS application complying
Suitability for the LC-MS complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR