Zinc oxide ≥99.0% (by complexometric analysis), Puriss. p.a. ACS, Fluka™

Supplier: Honeywell Research Chemicals

96479-500G
BJ96479-1KGEA 158.26 USD
BJ96479-1KG BJ96479-250G BJ96479-500G
Zinc oxide ≥99.0% (by complexometric analysis), Puriss. p.a. ACS, Fluka™
Zinc oxide
Formula: ZnO
Melting Pt: 1.975 °C
Density: 5.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011300
CAS Number: 1314-13-2
EINECS: 215-222-5

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
Insoluble in acid Max. 0.01 %
Alkalinely reacting substances complying
Arsenic (As) Max. 1 ppm
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 10 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 20 ppm
Chloride (Cl) Max. 10 ppm
Nitrate (NO₃) Max. 30 ppm
Total S (as SO₄) Max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR