p-Xylene ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: 1,4-Dimethylbenzene

95682-2.5L
BJ95682-2.5LEA 447.05 USD
BJ95682-2.5L
p-Xylene ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™
p-Xylene

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.


Beilstein Number: 1901563
RTECS Number: ZE2625000
UNSPSC Code: 12190000

Formula: C₆H₄(CH₃)₂
MW: 106.17 g/mol
Boiling Pt: 137…139 °C (1.013 hPa)
Melting Pt: 12 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00008556
CAS Number: 106-42-3
EINECS: 203-396-5
UN: 1307
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8b04563a5941cefd103ebd3048bcf8486f2bdac&vid=e5db83ad1f0cb6a583610c58cea291dcbe1be1f3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 99.0 %
Refractive index (n 20/D) 1.494 - 1.496
Non-volatile matter Max. 0.005 %
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
o-Xylene (GC) Max. 0.5 %
m-Xylole (GC) Max. 0.5 %
Sum of the isomers (GC) Max. 0.5 %
Benzene (GC) Max. 1.0 %
Ethyl benzene (GC) Max. 1.0 %
Toluene (GC) Max. 0.5 %
Refractive Index n20/D 1.495

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR