TraceSELECT™ Fluka™ Cesium Chloride - Lanthanum Chloride Buffer, Honeywell Reseach Chemicals
Supplier: Honeywell Research Chemicals
95164-250ML
BJ95164-250MLEA 434.32 USD
BJ95164-250ML
TraceSELECT™ Fluka™ Cesium Chloride - Lanthanum Chloride Buffer, Honeywell Reseach Chemicals
Buffers Ionisation Buffers
TraceSELECT™, for flame atomic absorption spectrometry.
  • Packaged in high-quality containers appropriate for each product
  • The reagents are produced and bottled under clean-room conditionsions
Lanthanum chloride (LaCl3) 99.0 - 101.0 g/l
Cesium chloride (Cscl) 99.0 - 101.0 g/l
Density (D 20/4) 1.152 - 1.176
Calcium (Ca) max. 0.5 ppm