Formic acid ∼98% (by titrimetric analysis) for mass spectrometry, Fluka™

Supplier: Honeywell Research Chemicals
Danger

94318-250ML
BJ94318-250MLEA 166 USD
BJ94318-250ML
Formic acid ∼98% (by titrimetric analysis) for mass spectrometry, Fluka™
Formic acid
Formula: HCO₂H
Boiling Pt: 100.4 °C (1.013 hPa)
Melting Pt: 4 °C
Density: 1.22 g/cm³ (20 °C)
Flash Pt: 49.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00003297
CAS Number: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8,II

Order Now

Specification Test Results

Assay Min. 97.5 %
Suitability for the LC-MS complying
Water (Karl Fischer) Max. 2.5 %
Non-volatile matter Max. 20 ppm
APHA Max. 15
Silver (Ag) Max. 0.02 ppm
Aluminium (Al) Max. 0.05 ppm
Barium (Ba) Max. 0.05 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.2 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.05 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.2 ppm
Potassium (K) Max. 0.1 ppm
Lithium (Li) Max. 0.02 ppm
Magnesium (Mg) Max. 0.5 ppm
Manganese (Mn) Max. 0.05 ppm
Molybdenum (Mo) Max. 0.02 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.05 ppm
Lead (Pb) Max. 0.02 ppm
Strontium (Sr) Max. 0.02 ppm
Thallium (Tl) Max. 0.05 ppm
Zinc (Zn) Max. 0.05 ppm
Chloride (Cl) Max. 5 ppm
Sulfate (SO₄) Max. 5 ppm
Sulfite (SO₃) Max. 10 ppm
Ammonium (NH₄) Max. 10 ppm
Appearance of the solution complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR