Citric acid monohydrate ≥99.9998% (metals basis), TraceSELECT™, Fluka™

Supplier: Honeywell Research Chemicals
Warning

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid

94068-100G
BJ94068-100GEA 77 USD
BJ94068-100G
Citric acid monohydrate ≥99.9998% (metals basis), TraceSELECT™, Fluka™
Citric acid monohydrate

TraceSELECT™ Ultra and TraceSELECT


  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

Formula: C₆H₈O₇.H₂O
MW: 210.14 g/mol
Melting Pt: 135…152 °C
Density: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149972
CAS Number: 5949-29-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3160681f96abd248f56672a96b72cc5f6e9d89a2&vid=c4db52c32c630d2c9876e60fc3e9e70a117ea0d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5 - 101.0 %
Silver (Ag) Max. 0.01 ppm
Aluminium (Al) Max. 0.05 ppm
Arsenic (As) Max. 0.01 ppm
Gold (Au) Max. 0.01 ppm
Barium (Ba) Max. 0.05 ppm
Beryllium (Be) Max. 0.01 ppm
Bismuth (Bi) Max. 0.01 ppm
Calcium (Ca) Max. 0.1 ppm
Cadmium (Cd) Max. 0.01 ppm
Cerium (Ce) Max. 0.01 ppm
Cobalt (Co) Max. 0.01 ppm
Chromium (Cr) Max. 0.05 ppm
Cesium (Cs) Max. 0.01 ppm
Copper (Cu) Max. 0.01 ppm
Dysprosium (Dy) Max. 0.01 ppm
Erbium (Er) Max. 0.01 ppm
Europium (Eu) Max. 0.01 ppm
Iron (Fe) Max. 0.1 ppm
Gadolinium (Gd) Max. 0.01 ppm
Germanium (Ge) Max. 0.01 ppm
Hafnium (Hf) Max. 0.01 ppm
Mercury (Hg) Max. 5 ppb
Holmium (Ho) Max. 0.01 ppm
Indium (In) Max. 0.01 ppm
Iridium (Ir) Max. 0.01 ppm
Potassium (K) Max. 0.1 ppm
Lanthanum (La) Max. 0.01 ppm
Lithium (Li) Max. 0.01 ppm
Lutetium (Lu) Max. 0.01 ppm
Magnesium (Mg) Max. 0.05 ppm
Manganese (Mn) Max. 0.01 ppm
Molybdenum (Mo) Max. 0.01 ppm
Sodium (Na) Max. 2 ppm
Niobium (nb) Max. 0.01 ppm
Neodymium (nd) Max. 0.01 ppm
Nickel (Ni) Max. 0.01 ppm
Lead (Pb) Max. 0.05 ppm
Palladium (Pd) Max. 0.01 ppm
Praseodymium (Pr) Max. 0.01 ppm
Platinum (Pt) Max. 0.01 ppm
Rubidium (Rb) Max. 0.01 ppm
Rhodium (Rh) Max. 0.01 ppm
Ruthenium (Ru) Max. 0.01 ppm
Antimony (Sb) Max. 0.01 ppm
Scandium (Sc) Max. 0.05 ppm
Selenium (Se) Max. 0.05 ppm
Samarium (Sm) Max. 0.01 ppm
Tin (Sn) Max. 0.01 ppm
Strontium (Sr) Max. 0.01 ppm
Tantalum (Ta) Max. 0.01 ppm
Terbium (Tb) Max. 0.01 ppm
Tellurium (Te) Max. 0.01 ppm
Thorium (Th) Max. 0.01 ppm
Titanium (Ti) Max. 0.05 ppm
Thallium (Tl) Max. 0.01 ppm
Thulium (Tm) Max. 0.01 ppm
Uran (U) Max. 0.01 ppm
Vanadium (V) Max. 0.01 ppm
Tungsten (W) Max. 0.01 ppm
Yttrium (Y) Max. 0.01 ppm
Ytterbium (Yb) Max. 0.01 ppm
Zinc (Zn) Max. 0.01 ppm
Zirconium (Zr) Max. 0.01 ppm
Bromide (Br) Max. 2000 ppb
Chloride (Cl) Max. 10000 ppb
Fluoride (F) Max. 2000 ppb
Iodide (I) Max. 2000 ppb
Nitrite (NO₂) Max. 2000 ppb
Nitrate (NO₃) Max. 10000 ppb
Phosphate (PO₄) Max. 2 ppm
Sulfate (SO₄) Max. 10000 ppb
Acetate (C₂H₃O₂) Max. 5000 ppb
Formate (CHO₂) Max. 5000 ppb
Glycolate (C₂H₃O₃) Max. 5000 ppb
Oxalate (C₂O₄) Max. 5000 ppb
Total N Max. 5 ppm
Tartrate Max. 0.2 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR