Non-Polar Silica Based SPE Columns, Honeywell Burdick & Jackson
Supplier: Honeywell Research Chemicals
9006 9034 9000 9004 9008 9010 9016 9018 9014 9002 9032 9012 9009
BJ9006PK 477 USD
BJ9006 BJ9034 BJ9000 BJ9004 BJ9008 BJ9010 BJ9016 BJ9018 BJ9014 BJ9002 BJ9032 BJ9012 BJ-9009
Non-Polar Silica Based SPE Columns, Honeywell Burdick & Jackson
Chromatography Columns
These SPE columns are made from medical-grade polypropylene with PE frits.
  • Wide selection of support materials and multiple cartridge/column sizes
  • Every stationary phase batch tested to ensure lot-to-lot reproducibility
  • Reliable quality
All columns are reversed phase and should be preconditioned with a polar solvent with the elution solvent being non-polar.