Tetrahydroxyquinone dihydrate indicator for titration (Ba, SO4), Fluka™

Supplier: Honeywell Research Chemicals
Warning

87510-5G
BJ87510-5GEA 443.56 USD
BJ87510-5G
Tetrahydroxyquinone dihydrate indicator for titration (Ba, SO4), Fluka™
Tetrahydroxyquinone dihydrate
Formula: C₆H₄O₆·₂H₂O
Storage Temperature: Ambient
MDL Number: MFCD00001597
CAS Number: 5676-48-2

Order Now

Specification Test Results

Ignition Residue ≤0.5%
Identity (IR) complying
Assay (calculated to the dried substance) 98.5 - 101.5 %
Colour of the solution complying
Clearness of the solution complying
Residue on ignition Max. 0.5 %
Application test complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR