Nitric acid fuming ≥98% (by titrimetric analysis), Purum p.a.

Supplier: Honeywell Research Chemicals
Danger

84392-500ML
BJ84392-500MLEA 299.16 USD
BJ84392-500ML
Nitric acid fuming ≥98% (by titrimetric analysis), Purum p.a.
Nitric acid
Formula: HNO₃
Boiling Pt: 86 °C
Melting Pt: –42 °C
Density: 1.52 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2032
ADR: 8,I
REACH: 01-2119487297-23
Merck Index: 13,06610

Order Now

Specification Test Results

Assay Min. 98 %
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 10 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Total S (as SO₄) Max. 150 ppm
Chloride (Cl) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR