Methanol : Water (60:40)%, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

837-4
BJ837-4CS 489.09 USD
BJ837-4
Methanol : Water (60:40)%, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Methanol (15 - < 30%)

LabReady™ Solvent Blends


  • Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
  • Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
  • Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
  • Honeywell's high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk

Formula: CH₄O/H₂O
Boiling Pt: 82.78 °C
Density: 0.9078 g/cm³ (20 °C)
Flash Pt: 18.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
UN: 1230
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR