Mercury(II) bromide ≥99.0% (by precipitation titration), Puriss. p.a. ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger

83353-50G
BJ83353-50GEA 205.89 USD
BJ83353-50G
Mercury(II) bromide ≥99.0% (by precipitation titration), Puriss. p.a. ACS, Fluka™
Mercury (II) bromide
Formula: Br₂Hg
Boiling Pt: 322 °C
Melting Pt: 237 °C
Density: 6.1 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011040
CAS Number: 7789-47-1
EINECS: 232-169-3
UN: 1634
ADR: 6.1,II

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
Residue on ignition a. reducing with HCOOH (SO₄)* Max. 0.02 %
Insoluble in methanol Max. 0.05 %
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Chloride (Cl) Max. 2000 ppm
Sulfate (SO₄) Max. 50 ppm
Ignition Residue (after reduction) ≤0.02%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR