Polyvinyl alcohol protective colloid for titrations, Fluka™

Supplier: Honeywell Research Chemicals
Warning

Synonyms: Poly(vinyl alcohol), PVOH, PVA, Mowiol®

81366-100G
BJ81366-100GEA 185.38 USD
BJ81366-100G
Polyvinyl alcohol protective colloid for titrations, Fluka™
Polyvinyl alcohol
Formula: [-CH₂CH(OH)-]n
Melting Pt: 200 °C
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now

Specification Test Results

Appearance complying
Identity (IR) complying
Colour of the solution complying
Clearness of the solution complying
pH (0,2 %, 25 °C) 4.5 - 7.4
Chloride (Cl) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR