Piperidine ≥99.0% (by GC, titration analysis), Puriss. p.a., Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

80640-1L
BJ80640-1LEA 552.7 USD
BJ80640-1L BJ80640-500ML
Piperidine ≥99.0% (by GC, titration analysis), Puriss. p.a., Riedel-de Haën™
Piperidine

Beilstein Number: 102438
RTECS Number: TM3500000
UNSPSC Code: 12190000

Formula: C₅H₁₁N
Boiling Pt: 10.3 %(V)
Melting Pt: -10 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: 16 °C
Storage Temperature: Ambient
MDL Number: MFCD00005979
CAS Number: 110-89-4
EINECS: 203-813-0
UN: 2401
ADR: 8,I

Order Now

Specification Test Results

Refractive Index n20/D 1.452 (lit.)
Identity (IR) complying
Assay (GC) Min. 99.5 %
Refractive index (n 20/D) 1.452 - 1.454
Non-volatile matter Max. 0.02 %
Water (Karl Fischer) Max. 0.3 %
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Tin (Sn) Max. 0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR