n-Octane ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

74821-500ML
BJ74821-500MLEA 373.22 USD
BJ74821-500ML
n-Octane ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™
n-Octane

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.


Beilstein Number: 1696875
RTECS Number: RG8400000
UNSPSC Code: 12190000

Formula: H₃C(CH₂)₆CH₃
MW: 114.23 g/mol
Boiling Pt: 6.5 %(V)
Melting Pt: –57 °C
Density: 0.703 g/cm³ (25 °C)
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00009556
CAS Number: 111-65-9
EINECS: 203-892-1
UN: 1262
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68adf890d68728825d4a2fa4b032700b93f710b8&vid=b00a2925902bc425c80f8ebb3f0e7b456c800ae3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (NMR) complying
Assay (GC) Min. 99.0 %
Refractive index (n 20/D) 1.397 - 1.399
Non-volatile matter Max. 0.002 %
Water (Karl Fischer) Max. 0.02 %
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
Explosion Limit 6.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR