Lithium chloride, anhydrous ≥99% ACS, free-flowing, Fluka™

Supplier: Honeywell Research Chemicals
Warning

746460-6X500G
BJ746460-500GCS 1316.24 USD
BJ746460-500G
Lithium chloride, anhydrous ≥99% ACS, free-flowing, Fluka™
Lithium chloride
Formula: LiCl
Boiling Pt: 1.360 °C (1.013 hPa)
Melting Pt: 605 °C
Density: 2.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
EINECS: 231-212-3

Order Now

Specification Test Results

Assay Min. 99.0 %
Water insoluble matter Max. 0.01 %
Loss on drying (105°C, 2 h) Max. 1.0 %
Titrable Base Max. 0.008 meq/g
Barium (Ba) Max. 0.003 %
Calcium (Ca) Max. 0.01 %
Iron (Fe) Max. 0.001 %
Potassium (K) Max. 0.01 %
Sodium (Na) Max. 0.20 %
Heavy metals (ICP) ACS Max. 0.002 %
Nitrate (NO₃) Max. 0.001 %
Sulfate (SO₄) Max. 0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR