Chromeazurol S (Mordant blue 29) indicator for metal titration, Fluka™

Supplier: Honeywell Research Chemicals

72687-25G
BJ72687-25GEA 180.72 USD
BJ72687-25G
Chromeazurol S (Mordant blue 29) indicator for metal titration, Fluka™
Chromeazurol S (Mordant blue 29)
Formula: C₂₃H₁₆Cl₂O₉S
Storage Temperature: Ambient
MDL Number: MFCD00001615
CAS Number: 1667-99-8
EINECS: 216-787-0

Order Now

Specification Test Results

Identity (IR) complying
Extinction maximum at 424 - 436 nm
Molar absorbancy index Min. 6000


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR