Sodium sulfate decahydrate ≥99.0% (by titrimetric analysis on dry substance), crystallised, Puriss. p.a., Fluka™

Supplier: Honeywell Research Chemicals

71970-1KG
BJ71970-1KGEA 92 USD
BJ71970-1KG
Sodium sulfate decahydrate ≥99.0% (by titrimetric analysis on dry substance), crystallised, Puriss. p.a., Fluka™
Formula: Na₂SO₄·10H₂O
Melting Pt: 32.4 °C
Density: 1.46 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149189
CAS Number: 7727-73-3

Order Now

Specification Test Results

Assay (calculated to the dried substance) 99.0 - 101.0 %
pH (5 %, 20°C) 5.0 - 8.0
Appearance of the solution complying
Loss on drying (130°C) 53 - 57 %
Calcium (Ca) Max. 20 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 20 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Chloride (Cl) Max. 5 ppm
Phosphate (PO₄) Max. 10 ppm
Total N Max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR