Sodium sulfate, anhydrous ≥99.0% (by titrimetric analysis), Purum p.a.

Supplier: Honeywell Research Chemicals

71962-5KG
BJ71962-5KGEA 510.15 USD
BJ71962-5KG
Sodium sulfate, anhydrous ≥99.0% (by titrimetric analysis), Purum p.a.
Sodium sulphate

  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Formula: Na₂SO₄
Melting Pt: 884 °C
Density: 2.68 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003504
CAS Number: 7757-82-6
EINECS: 231-820-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99.0 %
Loss on drying (130°C) Max. 1 %
pH (5 %, 20°C) 5.0 - 7.5
Calcium (Ca) Max. 0.005 %
Cadmium (Cd) Max. 0.005 %
Cobalt (Co) Max. 0.005 %
Copper (Cu) Max. 0.005 %
Iron (Fe) Max. 0.005 %
Potassium (K) Max. 0.01 %
Nickel (Ni) Max. 0.005 %
Lead (Pb) Max. 0.005 %
Zinc (Zn) Max. 0.005 %
Chloride (Cl) Max. 0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR