Sodium selenate ≥98.0% (by titrimetric analysis), Purum p.a.

Supplier: Honeywell Research Chemicals
Danger

71948-500G
BJ71948-500GEA 1231.2 USD
BJ71948-500G BJ71948-100G
Sodium selenate ≥98.0% (by titrimetric analysis), Purum p.a.
Sodium selenate

  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Formula: Na₂O₄Se
Density: 3.213 g/cm³ (17.5 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003490
CAS Number: 13410-01-0
EINECS: 236-501-8
UN: 2630
ADR: 6.1,I

Order Now

Specification Test Results

Assay 98.0 - 102.0 %
Calcium (Ca) Max. 200 ppm
Cadmium (Cd) Max. 50 ppm
Cobalt (Co) Max. 50 ppm
Copper (Cu) Max. 50 ppm
Iron (Fe) Max. 50 ppm
Potassium (K) Max. 1000 ppm
Nickel (Ni) Max. 50 ppm
Lead (Pb) Max. 50 ppm
Zinc (Zn) Max. 50 ppm
Chloride (Cl) Max. 1000 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR