Sodium hypochlorite ∼10% (RT) in aqueous solution, Purum, Fluka™

Supplier: Honeywell Research Chemicals
Danger

71696-10L
BJ71696-10LEA 357 USD
BJ71696-10L
Sodium hypochlorite ∼10% (RT) in aqueous solution, Purum, Fluka™
Sodium hypochlorite
Formula: ClNaO
Boiling Pt: 111 °C
Melting Pt: –30…–20 °C
Density: 1.206 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011120
CAS Number: 7681-52-9
EINECS: 231-668-3
UN: 1791
ADR: 8,II

Order Now

Specification Test Results

Assay of active chlorine 8.0 - 15.0 %
pH (direct) 11.5 - 13.5
Density (D 20/4) 1.21 - 1.26


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR