Sodium hydroxide ≥98.0% (by titrimetric analysis), pellets, Puriss. p.a. ACS (max. 0.02% K), Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

71690-1KG 71690-500G 71690-5KG
BJ71690-1KGEA 188.94 USD
BJ71690-1KG BJ71690-500G BJ71690-5KG
Sodium hydroxide ≥98.0% (by titrimetric analysis), pellets, Puriss. p.a. ACS (max. 0.02% K), Fluka™
Sodium hydroxide
Formula: NaOH
Boiling Pt: 1.390 °C (1.013 hPa)
Melting Pt: 319 °C
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II

Order Now

Specification Test Results

Assay Min. 98.0 %
Assay of Na₂CO₃ Max. 1.0 %
Silver (Ag) Max. 0.0005 %
Aluminium (Al) Max. 0.0005 %
Barium (Ba) Max. 0.0005 %
Bismuth (Bi) Max. 0.0005 %
Calcium (Ca) Max. 0.0005 %
Cadmium (Cd) Max. 0.0005 %
Cobalt (Co) Max. 0.0005 %
Chromium (Cr) Max. 0.0005 %
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 0.0005 %
Mercury (Hg) Max. 0.00001 %
Potassium (K) Max. 0.02 %
Lithium (Li) Max. 0.0005 %
Magnesium (Mg) Max. 0.0005 %
Manganese (Mn) Max. 0.0005 %
Molybdenum (Mo) Max. 0.0005 %
Nickel (Ni) Max. 0.0005 %
Lead (Pb) Max. 0.0005 %
Strontium (Sr) Max. 0.0005 %
Thallium (Tl) Max. 0.0005 %
Zinc (Zn) Max. 0.0005 %
Heavy metals (as Ag) Max. 0.002 %
Chloride (Cl) Max. 0.005 %
Total N Max. 0.0003 %
Phosphate (PO₄) Max. 0.0005 %
Silicate (as SiO₂) Max. 0.0005 %
Sulfate (SO₄) Max. 0.003 %
Vapor Pressure 3 mmHg ( 37 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR