Sodium hydrogen selenite ≥96.0% (RT), Purum p.a.

Supplier: Honeywell Research Chemicals
Danger

71658-100G
BJ71658-100GEA 307.62 USD
BJ71658-100G
Sodium hydrogen selenite ≥96.0% (RT), Purum p.a.
Sodium hydrogen selenite
Formula: HNaO₃Se
Storage Temperature: Ambient
MDL Number: MFCD00003525
CAS Number: 7782-82-3
EINECS: 231-966-3
UN: 2630
ADR: 6.1,I

Order Now

Specification Test Results

Assay 96.0 - 104.0 %
pH (5 %, 20°C) 5.4 - 7.2
Colour of the solution complying
Clearness of the solution complying
Calcium (Ca) Max. 100 ppm
Cadmium (Cd) Max. 50 ppm
Cobalt (Co) Max. 50 ppm
Copper (Cu) Max. 100 ppm
Iron (Fe) Max. 50 ppm
Potassium (K) Max. 100 ppm
Nickel (Ni) Max. 50 ppm
Lead (Pb) Max. 300 ppm
Zinc (Zn) Max. 100 ppm
Chloride (Cl) Max. 100 ppm
Selenate and sulfate (as SO₄) Max. 1500 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR