Disodium hydrogen phosphate dihydrate ≥98.5% for HPLC, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: sec-Sodium phosphate, Sodium monohydrogen phosphate dihydrate, Sodium hydrogenphosphate dihydrate, di-Sodium hydrogen orthophosphate dihydrate, Sodium phosphate dibasic dihydrate, Disodium phosphate, di-Sodium hydrogen phosphate dihydrate, Sorensen's salt

71633-250G
BJ71633-250GEA 149 USD
BJ71633-250G
Disodium hydrogen phosphate dihydrate ≥98.5% for HPLC, Fluka™
di-Sodium hydrogen phosphate dihydrate
Formula: Na₂HPO₄·2H₂O
MW: 177.99 g/mol
Melting Pt: 450 °C
Density: 2.07 g/cm³ (16 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149182
CAS Number: 10028-24-7

Order Now

Specification Test Results

Assay 98.5 - 102.0 %
pH (10 %) 8.8 - 9.4
Colour of the solution complying
Turbidity of solution complying
Absorbance 500 nm Max. 0.02
Absorbance at 320 nm Max. 0.02
Absorbance 280 nm Max. 0.04
Absorbance at 260 nm Max. 0.06
Absorbance at 230 nm Max. 0.1

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR