Disodium hydrogen phosphate, anhydrous ≥99.999% (metals basis), TraceSELECT™ for trace analysis, Fluka™
Supplier: Honeywell Research Chemicals
71629-100G
BJ71629-100GEA
667.75
USD
BJ71629-100G
Disodium hydrogen phosphate, anhydrous ≥99.999% (metals basis), TraceSELECT™ for trace analysis, Fluka™
di-Sodium hydrogen phosphate
TraceSELECT™ Ultra and TraceSELECT
- ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions
Specification Test Results
Arsenic (As) | Max. 0.1 ppm |
Barium (Ba) | Max. 1 ppm |
Calcium (Ca) | Max. 0.5 ppm |
Cadmium (Cd) | Max. 0.01 ppm |
Cerium (Ce) | Max. 0.01 ppm |
Cobalt (Co) | Max. 0.01 ppm |
Cesium (Cs) | Max. 5 ppm |
Copper (Cu) | Max. 0.01 ppm |
Europium (Eu) | Max. 0.01 ppm |
Iron (Fe) | Max. 0.05 ppm |
Mercury (Hg) | Max. 5 ppb |
Iridium (Ir) | Max. 2 ppm |
Potassium (K) | Max. 2 ppm |
Lanthanum (La) | Max. 0.01 ppm |
Lithium (Li) | Max. 1 ppm |
Magnesium (Mg) | Max. 0.1 ppm |
Manganese (Mn) | Max. 0.01 ppm |
Nickel (Ni) | Max. 0.01 ppm |
Lead (Pb) | Max. 0.01 ppm |
Palladium (Pd) | Max. 0.5 ppm |
Platinum (Pt) | Max. 0.5 ppm |
Rubidium (Rb) | Max. 5 ppm |
Rhodium (Rh) | Max. 0.5 ppm |
Ruthenium (Ru) | Max. 0.5 ppm |
Antimony (Sb) | Max. 0.05 ppm |
Selenium (Se) | Max. 0.01 ppm |
Samarium (Sm) | Max. 0.01 ppm |
Strontium (Sr) | Max. 0.5 ppm |
Thallium (Tl) | Max. 0.01 ppm |
Vanadium (V) | Max. 0.1 ppm |
Yttrium (Y) | Max. 0.01 ppm |
Ytterbium (Yb) | Max. 0.01 ppm |
Zinc (Zn) | Max. 0.05 ppm |
Chloride (Cl) | Max. 5 ppm |
Sulfate (SO₄) | Max. 30 ppm |
Total nitrogen (as N) | Max. 10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...