Sodium dihydrogen phosphate, anhydrous ≥99.999% (metals basis), TraceSELECT™ for trace analysis, Fluka™

Supplier: Honeywell Research Chemicals

71492-100G
BJ71492-100GEA 508.49 USD
BJ71492-100G BJ71492-25G
Sodium dihydrogen phosphate, anhydrous ≥99.999% (metals basis), TraceSELECT™ for trace analysis, Fluka™
Sodium dihydrogen phosphate

TraceSELECT™ Ultra and TraceSELECT


  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

Formula: NaH₂PO₄
Melting Pt: 650 °C
Density: 2.36 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003527
CAS Number: 7558-80-7
EINECS: 231-449-2

Order Now

Specification Test Results

Arsenic (As) Max. 0.01 ppm
Barium (Ba) Max. 1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.01 ppm
Cerium (Ce) Max. 0.01 ppm
Cobalt (Co) Max. 0.01 ppm
Cesium (Cs) Max. 5 ppm
Copper (Cu) Max. 0.01 ppm
Europium (Eu) Max. 0.01 ppm
Iron (Fe) Max. 0.05 ppm
Mercury (Hg) Max. 5 ppb
Iridium (Ir) Max. 2 ppm
Potassium (K) Max. 5 ppm
Lanthanum (La) Max. 0.01 ppm
Lithium (Li) Max. 1 ppm
Magnesium (Mg) Max. 0.5 ppm
Manganese (Mn) Max. 0.01 ppm
Nickel (Ni) Max. 0.01 ppm
Lead (Pb) Max. 0.01 ppm
Palladium (Pd) Max. 0.5 ppm
Platinum (Pt) Max. 0.5 ppm
Rubidium (Rb) Max. 5 ppm
Rhodium (Rh) Max. 0.5 ppm
Ruthenium (Ru) Max. 0.5 ppm
Antimony (Sb) Max. 0.2 ppm
Selenium (Se) Max. 0.01 ppm
Samarium (Sm) Max. 0.01 ppm
Strontium (Sr) Max. 0.5 ppm
Thallium (Tl) Max. 0.01 ppm
Vanadium (V) Max. 0.1 ppm
Yttrium (Y) Max. 0.01 ppm
Ytterbium (Yb) Max. 0.01 ppm
Zinc (Zn) Max. 0.01 ppm
Chloride (Cl) Max. 5 ppm
Sulfate (SO₄) Max. 30 ppm
Total nitrogen (as N) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR