Sodium hydroxide 1 M NaOH (1N) ≥99.5% volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

71463-1L
BJ71463-1LEA 52 USD
BJ71463-1L
Sodium hydroxide 1 M NaOH (1N) ≥99.5% volumetric solution, Fluka™
Sodium hydroxide
Formula: NaOH
Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 1.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Calibrated Against potassium hydrogen phthalate
Assay 0.998 - 1.002 mol/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR