Sodium hydroxide 0.1 M (0.1 N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Warning

71395-1L
BJ71395-1LEA 32 USD
BJ71395-1L
Sodium hydroxide 0.1 M (0.1 N) volumetric solution, Fluka™
Sodium hydroxide
Formula: NaOH
Boiling Pt: 100 °C (1.013 hPa)
Melting Pt: 319 °C
Density: 1.000 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 231-791-2
UN: 1824
ADR: 8,III

Order Now

Specification Test Results

Assay 0.0998 - 0.1002 mol/l
Calibrated Against potassium hydrogen phthalate


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR