Sodium chloride 1.0 M NaCl (1.0N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals

71394-1L
BJ71394-1LEA 163.53 USD
BJ71394-1L
Sodium chloride 1.0 M NaCl (1.0N) volumetric solution, Fluka™
Sodium chloride
Formula: NaCl
Density: 1.00 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5

Order Now

Specification Test Results

Density (D 20/4) 1.030 - 1.050
Factor 0.995 - 1.005


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR