Sodium carbonate, anhydrous ≥99.999% (metals basis), TraceSELECT™ for trace analysis, Fluka™
Supplier: Honeywell Research Chemicals
|
Warning
|
Synonyms:
Soda, Carbonic acid sodium salt, Calcined soda, Disodium carbonate, Carbonic acid disodium salt, Na-X, Bisodium carbonate, Carbonic acid, disodium salt, Soda ash
71347-100G
71347-25G
BJ71347-100GEA
446
USD
BJ71347-100G
BJ71347-25G
Sodium carbonate, anhydrous ≥99.999% (metals basis), TraceSELECT™ for trace analysis, Fluka™
Sodium carbonate
TraceSELECT™ Ultra and TraceSELECT
- ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions
Formula:
Na₂CO₃ MW: 105.99 g/mol Boiling Pt: 1600 Melting Pt: 854 Density: 2.53 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003494 CAS Number: 497-19-8 EINECS: 207-838-8 Merck Index: 14,08596 |
|
Specification Test Results
Purity based on trace metals analysis | Min. 99.999 % |
Aluminium (Al) | Max. 0.05 ppm |
Arsenic (As) | Max. 0.01 ppm |
Barium (Ba) | Max. 5 ppm |
Calcium (Ca) | Max. 0.1 ppm |
Cadmium (Cd) | Max. 0.005 ppm |
Cerium (Ce) | Max. 0.01 ppm |
Cobalt (Co) | Max. 0.005 ppm |
Chromium (Cr) | Max. 0.01 ppm |
Copper (Cu) | Max. 0.005 ppm |
Europium (Eu) | Max. 0.01 ppm |
Iron (Fe) | Max. 0.05 ppm |
Mercury (Hg) | Max. 5 ppb |
Iridium (Ir) | Max. 0.5 ppm |
Potassium (K) | Max. 1 ppm |
Lithium (Li) | Max. 0.5 ppm |
Lanthanum (La) | Max. 0.01 ppm |
Magnesium (Mg) | Max. 0.1 ppm |
Manganese (Mn) | Max. 0.01 ppm |
Nickel (Ni) | Max. 0.01 ppm |
Lead (Pb) | Max. 0.01 ppm |
Palladium (Pd) | Max. 0.5 ppm |
Platinum (Pt) | Max. 0.5 ppm |
Rhodium (Rh) | Max. 0.5 ppm |
Ruthenium (Ru) | Max. 0.5 ppm |
Antimony (Sb) | Max. 0.01 ppm |
Scandium (Sc) | Max. 0.01 ppm |
Selenium (Se) | Max. 0.01 ppm |
Samarium (Sm) | Max. 0.01 ppm |
Strontium (Sr) | Max. 0.1 ppm |
Thallium (Tl) | Max. 0.01 ppm |
Vanadium (V) | Max. 0.1 ppm |
Yttrium (Y) | Max. 0.01 ppm |
Ytterbium (Yb) | Max. 0.01 ppm |
Zinc (Zn) | Max. 0.01 ppm |
Chloride (Cl) | Max. 10 ppm |
Total N | Max. 10 ppm |
Phosphate (PO₄) | Max. 5 ppm |
Sulfate (SO₄) | Max. 10 ppm |
Silicate (as SiO₂) | Max. 5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...