Sodium borohydride ≥99%, TraceSELECT™ for the determination of hydride formers by aas, Fluka™

Supplier: Honeywell Research Chemicals
Danger

71321-100G
BJ71321-100GEA 307 USD
BJ71321-100G BJ71321-25G
Sodium borohydride ≥99%, TraceSELECT™ for the determination of hydride formers by aas, Fluka™
Sodium borohydride

TraceSELECT™ Ultra and TraceSELECT


  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

Formula: NaBH₄
Boiling Pt: 400 °C (1.027 hPa)
Melting Pt: 360 °C
Density: 1.074 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003518
CAS Number: 16940-66-2
EINECS: 241-004-4
UN: 1426
ADR: 4.3,I

Order Now

Specification Test Results

Assay 98.5 - 101.5 %
Arsenic (As) Max. 0.05 ppm
Mercury (Hg) Max. 50 ppb
Antimony (Sb) Max. 0.05 ppm
Selenium (Se) Max. 0.05 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR