Acetic acid ≥99.7% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger

695092-6X500ML
BJ695092-500MLCS 313.69 USD
BJ695092-500ML
Acetic acid ≥99.7% ACS, Fluka™
Acetic acid
Formula: H₃CCOOH
Boiling Pt: 118 °C (1.013 hPa)
Melting Pt: 16 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance complying
Identity (IR) complying
Assay (GC) Min. 99.7 %
Non-volatile matter Max. 0.001 %
Titrable Base Max. 0.0004 meq/g
Iron (Fe) Max. 0.00002 %
Heavy metals (as Pb) Max. 0.00005 %
Chloride (Cl) Max. 0.0001 %
Sulfate (SO₄) Max. 0.0001 %
Acetic aNHydride (GC) Max. 0.01 %
KMnO₄ reducing substances complying
K₂Cr₂O₇ reducing matter complying
APHA Max. 10
Mixable with H₂O complying
Explosion Limit 16 %, 92 °F
Refractive Index n20/D 1.371 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR