Water with 0,05% (v/v) formic acid, CHROMASOLV™ LabReady™, Solvent Blend for HPLC, Riedel-de Haën™

Supplier: Honeywell Research Chemicals

679178-4X4L
BJ679178-4X4LCS 724.39 USD
BJ679178-4X4L
Water with 0,05% (v/v) formic acid, CHROMASOLV™ LabReady™, Solvent Blend for HPLC, Riedel-de Haën™
Water with 0,05% (v/v) formic acid

  • High purity chromatography solvents
  • Suitability tested for specified use from HPLC up to UHPLC-MS applications.


UNSPSC Code: 12190000

Boiling Pt: 100 °C
Density: 0.9986 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
EINECS: 231-791-2

Order Now

Specification Test Results

Contains 0.1 %(v/v) formic acid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR