Carbon disulfide ≥99.9% ACS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

676918-1L
BJ676918-1LEA 523 USD
BJ676918-1L
Carbon disulfide ≥99.9% ACS, Riedel-de Haën™
Carbon disulphide

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Formula: CS₂
MW: 76.14 g/mol
Boiling Pt: 46 °C (1.013 hPa)
Melting Pt: -76 °C
Density: 1.264 g/cm³ (20 °C)
Flash Pt: -30 °C
Storage Temperature: Ambient
MDL Number: MFCD00011321
CAS Number: 75-15-0
EINECS: 200-843-6
UN: 1131
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7fe0c906449b3b5461b1bac971c583c92c0b41c&vid=59e9028fbb4d1c2acb6f98101726e8ec520e0aa8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 99.90 %
Non-volatile matter Max. 0.002 %
Water (Karl Fischer) Max. 0.05 %
Sulfur dioxide (SO₂) complying
Hydrogen sulfide (H₂S) complying
APHA Max. 10
Autoignition Temperature 212 °F
Vapor Density 2.67 (vs air)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR