Petroleum spirit, 35…60 °C ACS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

673838-4X4L
BJ673838-4LCS 1114.41 USD
BJ673838-4L
Petroleum spirit, 35…60 °C ACS, Riedel-de Haën™
Petroleum spirit

Petroleum ethers are low boiling aliphatic fractions, which are derived from crude petroleum via fractional distillation. The boiling range is 40-60°C, which gives a high evaporation rate. Suitable for general laboratory use; Meets ACS General Use specifications.


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications

Boiling Pt: 5.9 %(V)
Melting Pt: –73 °C
Density: 0.64 g/cm³ (20 °C) (lit.)
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 232-453-7
UN: 1268
ADR: 3,II

Order Now

Specification Test Results

Autoignition Temperature 475 °F
Identity (IR) complying
Refractive index (n 20/D) 1.358 - 1.368
Non-volatile matter Max. 0.001 %
Boiling range 35,0 - 60,0 °C
Vapor Pressure 25.8 psi ( 55 °C)
Acidity complying
APHA Max. 10
Explosion Limit 8 %
Vapor Density 2.5 (vs air)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR