Triethylamine ≥99.5% (by GC) eluent additive, for LC-MS, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: (Diethylamino)ethane, TEA

65897-50ML
BJ65897-50MLEA 102.46 USD
BJ65897-50ML
Triethylamine ≥99.5% (by GC) eluent additive, for LC-MS, Fluka™
Triethylamine

Eluent additive for LC-MS.

Formula: (C₂H₅)₃N
Boiling Pt: 90 °C
Melting Pt: –115 °C
Density: 0.73 g/cm3 (20 °C)
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00009051
CAS Number: 121-44-8
EINECS: 204-469-4
UN: 1296
ADR: 3 (8),II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2b1f184e438ecb01f88a4d90ee46c0d4316859d&vid=1c68408a0eedf1bfdb02d1b1e2337d50d449cf9d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) min. 99.5 %
Refractive index (n 20/D) 1.400 - 1.402
Appearance complying
Non-volatile matter max. 0.003 %
Water (Karl Fischer) max. 0.5 %
Aluminium (Al) max. 0.2 ppm
Barium (Ba) max. 0.1 ppm
Bismuth (Bi) max. 0.1 ppm
Calcium (Ca) max. 0.5 ppm
Cadmium (Cd) max. 0.05 ppm
Cobalt (Co) max. 0.02 ppm
Chromium (Cr) max. 0.02 ppm
Copper (Cu) max. 0.02 ppm
Iron (Fe) max. 0.1 ppm
Potassium (K) max. 0.5 ppm
Lithium (Li) max. 0.1 ppm
Magnesium (Mg) max. 0.1 ppm
Manganese (Mn) max. 0.02 ppm
Molybdenum (Mo) max. 0.02 ppm
Sodium (Na) max. 0.5 ppm
Nickel (Ni) max. 0.02 ppm
Lead (Pb) max. 0.1 ppm
Strontium (Sr) max. 0.1 ppm
Zinc (Zn) max. 0.1 ppm
UV- cutoff 250.0 - 278.0 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR