tert-Butyl methyl ether ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

650560-1L
BJ650560-1LEA 213 USD
BJ650560-1L
tert-Butyl methyl ether ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
tert-Butyl methyl ether

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities

Formula: (CH₃)₃COCH₃
Boiling Pt: 55.3 °C (1.013 hPa)
Melting Pt: –109 °C
Density: 0.74 g/cm³ (20 °C)
Flash Pt: 8.5 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00008812
CAS Number: 1634-04-4
EINECS: 216-653-1
UN: 2398
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1712bc714ba0a06e0de37f7cf4e5117256f84425&vid=e1b314f829136075329c50f9e5a593c19ae28b4f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) Min. 99.85 %
IR spectrum complying
Non-volatile matter Max. 0.0001 %
Water (Karl Fischer) Max. 0.02 %
Absorbance at 400 nm Max. 0.005
Absorbance at 300 nm Max. 0.005
Absorbance at 250 nm Max. 0.10
Absorbance at 225 nm Max. 0.50
Absorbance at 210 nm Max. 1.0
Fluorescence (chinin) at 365 nm Max. 1.0 ppb
GC (as heptachlor epoxide) Max. 10 ng/l
Refractive Index n20/D 1.369 (lit.)
Autoignition Temperature 705 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR