n-Hexane ≥98.5% (mixture of isomers), CHROMASOLV™ Plus for HPLC, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

650544-200L
BJ650544-200LEA 5209.5 USD
BJ650544-200L
n-Hexane ≥98.5% (mixture of isomers), CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
n-Hexane
Formula: H₃C(CH₂)₄CH₃
Boiling Pt: 48…65 °C(1,013 hPa)
Melting Pt: –154 °C
Density: 0.66 g/cm³ (20 °C)
Flash Pt: –26 °C
Storage Temperature: Room Temperature
MDL Number: MFCD00009520
CAS Number: 110-54-3
UN: 1208
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay of hexane (all isomers, GC) min. 98.50 %
Assay of n-Hexane (GC) min. 60.0 %
Refractive index (n 20/D) 1.377 - 1.382
Non-volatile matter max. 0.0001 %
Water (Karl Fischer) max. 0.01 %
GC (as heptachlor epoxide) max. 10 ng/l
Absorbance at 300 nm max. 0.005
Absorbance at 275 nm max. 0.005
Absorbance at 250 nm max. 0.01
Absorbance at 225 nm max. 0.05
Absorbance at 195 nm max. 1.0
Fluorescence (chinin) at 365 nm max. 1.0 ppb


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR