Acetone ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

650501-4X4L
BJ650501-4LCS 1077 USD
BJ650501-4L BJ650501-1L
Acetone ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Acetone
Formula: CH₃COCH₃
Boiling Pt: 55…57 °C (1.013 hPa)
Melting Pt: –95 °C
Density: 0.79 g/cm³ (20 °C) (lit.)
Flash Pt: 14.3 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Refractive Index n20/D 1.359 (lit.)
Purity (GC) Min. 99.90 %
GC (as heptachlor epoxide) Max. 10 ng/l
Water (Karl Fischer) Max. 0.5 %
Non-volatile matter Max. 0.0003 %
IR spectrum complying
Absorbance at 400 nm Max. 0.005
Absorbance at 375 nm Max. 0.005
Absorbance at 350 nm Max. 0.01
Absorbance at 340 nm Max. 0.06
Absorbance at 330 nm Max. 1.0
Fluorescence (chinin) at 365 nm Max. 1.0 ppb
Explosion Limit 13.2 %
Vapor Pressure 184 mmHg ( 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR