2-Propanol ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Isopropyl rubbing alcohol, sec-Propanol, Isopropanol, 2-Hydroxypropane, Isopropyl Rubbing Alcohol, 2-Hydroxy propane, IPA, sec-Propyl Alcohol, Dimethyl carbinol, Propane, 2-hydroxy-, Isopropyl alcohol, Propan-2-ol
2-Propanol (Isopropyl alcohol, Isopropanol, IPA) is a colorless solvent and is the simplest example of a secondary alcohol. It is moderately polar and has a boiling point of 82 °C. 2-Propanol dissolves a wide range of non-polar compounds and evaporates quickly. Thus, it is used widely as a cleaning fluid, especially for dissolving oils. Suitable for HPLC , spectrophotometry, GC and residue analysis.
- For HPLC, spectrophotometry, and environmental analysis
- Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
- Low UV impurity profile
- Low impurity gradient LC baseline
- Very low levels of non-volatile impurities
Formula:
(CH₃)₂CHOH Boiling Pt: 77…83 °C (1.013 hPa) Melting Pt: –90 °C Density: 0.784…0.786 g/cm³ (20 °C) (lit.) Flash Pt: 12 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011674 CAS Number: 67-63-0 EINECS: 200-661-7 UN: 1219 ADR: 3,II |
|
Specification Test Results
IR spectrum | complying |
Assay (GC) | Min. 99.85 % |
Non-volatile matter | Max. 0.0001 % |
Water (Karl Fischer) | Max. 0.02 % |
Absorbance at 400 nm | Max. 0.005 |
Absorbance at 275 nm | Max. 0.005 |
Absorbance at 250 nm | Max. 0.30 |
Absorbance at 245 nm | max. 1.0 |
Fluorescence (chinin) at 365 nm | max. 0.5 ppb |
GC (as heptachlor epoxide) | max. 10 ng/l |
Vapor Density | 2.1 (vs air) |
Autoignition Temperature | 750 °F |
Explosion Limit | 2 % |
Vapor Pressure | 33 mmHg ( 20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...