Magnesium sulfate 1.0 M MgSO4 volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: MgSO4, Magnesium sulfate anhydrous, Magnesium sulphate anhydrous

63126-1L
BJ63126-1LEA 368.72 USD
BJ63126-1L
Magnesium sulfate 1.0 M MgSO4 volumetric solution, Fluka™
Magnesium sulphate
Formula: MgSO₄
MW: 120.37 g/mol
Boiling Pt: 100 °C
Melting Pt: 1127
Density: 1.10 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 7487-88-9
EINECS: 231-791-2
Merck Index: 12,05731
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Factor 0.995 - 1.005


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR