Magnesium chloride 1.0 M MgCl2 volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals

63020-1L
BJ63020-1LEA 54 USD
BJ63020-1L
Magnesium chloride 1.0 M MgCl2 volumetric solution, Fluka™
Magnesium chloride
Formula: MgCl₂
Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 1.08 g/cm³
Flash Pt: Does not flash
Storage Temperature: Ambient
MDL Number: MFCD00011106
CAS Number: 7786-30-3

Order Now

Specification Test Results

Assay 0.995 - 1.005 mol/l
Density (D 20/4) 1.070 - 1.090


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR