Copper(II) oxide ≥99.0% (RT), powder, Puriss. p.a., Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: Cupric oxide, Copper oxide, Copper monoxide granulated

61200-500G
BJ61200-100GEA 130.33 USD
BJ61200-100G BJ61200-500G
Copper(II) oxide ≥99.0% (RT), powder, Puriss. p.a., Fluka™
Copper (II) oxide
Formula: CuO
MW: 79.55 g/mol
Boiling Pt: 2000
Melting Pt: 1326
Density: 6.450 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010979
CAS Number: 1317-38-0
EINECS: 215-269-1
Merck Index: 13,02674

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
Insoluble in HCl Max. 200 ppm
Calcium (Ca) Max. 100 ppm
Cadmium (Cd) Max. 10 ppm
Cobalt (Co) Max. 10 ppm
Chromium (Cr) Max. 50 ppm
Iron (Fe) Max. 500 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 100 ppm
Manganese (Mn) Max. 10 ppm
Sodium (Na) Max. 500 ppm
Nickel (Ni) Max. 50 ppm
Lead (Pb) Max. 500 ppm
Zinc (Zn) Max. 100 ppm
Chloride (Cl) Max. 50 ppm
Total S (as SO₄) Max. 500 ppm
Total N Max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR